Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Abl Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB086164 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB086164 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB086164 Supplier Novus Biologicals Supplier No. NBP299737AF594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

c-Abl Polyclonal specifically detects c-Abl in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen c-Abl
Applications ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50 mM Sodium Borate
Gene Alias Abelson murine leukemia viral oncogene homolog 1, ABL, c-ABL, c-abl oncogene 1, non-receptor tyrosine kinase, c-abl oncogene 1, receptor tyrosine kinase, EC 2.7.10, EC 2.7.10.2, JTK7bcr/abl, p150bcr/c-abl oncogene protein, Proto-oncogene c-Abl, proto-oncogene tyrosine-protein kinase ABL1, tyrosine-protein kinase ABL1, v-abl, v-abl Abelson murine leukemia viral oncogene homolog 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant c-Abl (NP_009297.2; Pro137-Ser554).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, DNA Repair, Phospho Specific, Signal Transduction, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 25
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.