Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C. botulinum BoNT-E Light Chain Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | BoNT-E Light Chain |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
BoNT-E Light Chain Polyclonal specifically detects BoNT-E Light Chain in C. botulinum samples. It is validated for Western Blot.Specifications
BoNT-E Light Chain | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
C. botulinum | |
E. coli-derived recombinant C. botulinum BoNT-E Light Chain Pro2-Ile403 Accession # P30995 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects C. botulinum BoNT-E Light Chain in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with rBoNT-B LC is observed and less than 1% cross-reactivity with rBoNT-C HCR and rBoNT-D HCR is observed. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
P30995 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title