Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Fos Antibody (2H2), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB360545 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB360545 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB360545 Supplier Novus Biologicals Supplier No. NBP250037AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

c-Fos Monoclonal antibody specifically detects c-Fos in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, KnockDown

Specifications

Antigen c-Fos
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, KnockDown
Classification Monoclonal
Clone 2H2
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Knockout Validated
Formulation 50mM Sodium Borate
Gene Alias activator protein 1, AP-1, cellular oncogene c-fos, Cellular oncogene fos, C-FOS, FBJ murine osteosarcoma viral (v-fos) oncogene homolog (oncogene FOS), FBJ murine osteosarcoma viral oncogene homolog, FOS, Fos proto-oncogene, AP-1 trancription factor subunit, G0/G1 switch regulatory protein 7, G0S7, p55, proto-oncogene c-Fos
Host Species Mouse
Immunogen This c-Fos Antibody (2H2) was developed against full length recombinant human c-Fos protein expressed in and purified from E. coli. [UniProt# P01100]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cellular Markers, Hypoxia, Neuronal Cell Markers, Neuroscience, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 2353
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.