Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-jun Antibody (OTI2D11), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP271058G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271058G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271058G Supplier Novus Biologicals Supplier No. NBP271058G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

c-jun Monoclonal specifically detects c-jun in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen c-Jun
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2D11
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Activator protein 1, AP1, AP-1, c-Jun, enhancer-binding protein AP1, Jun activation domain binding protein, jun oncogene, jun proto-oncogene, Proto-oncogene c-Jun, transcription factor AP-1, V-jun avian sarcoma virus 17 oncogene homologp39, v-jun sarcoma virus 17 oncogene homolog, v-jun sarcoma virus 17 oncogene homolog (avian)
Gene Symbols JUN
Host Species Mouse
Immunogen Full length recombinant protein of human c-jun (NP_002219) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cancer, Cell Cycle and Replication, Hypoxia, Immunology, Innate Immunity, Phospho Specific, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 3725.0
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.