Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-jun Antibody (OTI3G2), PE, Novus Biologicals™
SDP

Catalog No. NBP271059PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271059PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271059PE Supplier Novus Biologicals Supplier No. NBP271059PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

c-jun Monoclonal specifically detects c-jun in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen c-Jun
Applications Flow Cytometry
Classification Monoclonal
Clone OTI3G2
Conjugate PE
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Activator protein 1, AP1, AP-1, c-Jun, enhancer-binding protein AP1, Jun activation domain binding protein, jun oncogene, jun proto-oncogene, Proto-oncogene c-Jun, transcription factor AP-1, V-jun avian sarcoma virus 17 oncogene homologp39, v-jun sarcoma virus 17 oncogene homolog, v-jun sarcoma virus 17 oncogene homolog (avian)
Gene Symbols JUN
Host Species Mouse
Immunogen Full length recombinant protein of human Jun (NP_002219) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cancer, Cell Cycle and Replication, Hypoxia, Immunology, Innate Immunity, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 3725.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.