Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-jun, p Thr91, p Thr93 Antibody (C-J 4C4/1), Biotin, Novus Biologicals™
SDP

Catalog No. NB1002748B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002748B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002748B Supplier Novus Biologicals Supplier No. NB1002748B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

c-Jun Monoclonal specifically detects c-Jun in Human, Rat samples. It is validated for ELISA, Western Blot.

Specifications

Antigen c-Jun
Applications ELISA, Western Blot
Classification Monoclonal
Clone C-J 4C4/1
Conjugate Biotin
Dilution Western Blot, ELISA
Formulation PBS with 0.05% Sodium Azide
Gene Alias Activator protein 1, AP1, AP-1, c-Jun, enhancer-binding protein AP1, Jun activation domain binding protein, jun oncogene, jun proto-oncogene, Proto-oncogene c-Jun, transcription factor AP-1, V-jun avian sarcoma virus 17 oncogene homologp39, v-jun sarcoma virus 17 oncogene homolog, v-jun sarcoma virus 17 oncogene homolog (avian)
Gene Symbols JUN
Host Species Mouse
Immunogen C-Jun phosphorylated at T91 and T93
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Cycle and Replication, Hypoxia, Immunology, Innate Immunity, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 3725
Target Species Human, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.