Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Maf Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB600267H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600267H 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600267H Supplier Novus Biologicals Supplier No. NB600267H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

c-Maf Polyclonal specifically detects c-Maf in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen c-Maf
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate HRP
Dilution Western Blot, Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias Avian musculoaponeurotic fibrosarcoma (MAF) protooncogene, c-MAF, c-maf proto-oncogene, MGC71685, Proto-oncogene c-Maf, T lymphocyte c-maf long form, transcription factor Maf, v-maf musculoaponeurotic fibrosarcoma (avian) oncogene homolog, V-maf musculoaponeurotic fibrosarcoma oncogene homolog, v-maf musculoaponeurotic fibrosarcoma oncogene homolog (avian)
Gene Symbols MAF
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 150 and 200 of mouse v-maf musculoaponeurotic fibrosarcoma oncogene homolog (c-Maf) using the numbering given in entry NP_001020748.2 (GeneID 17132).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4094
Target Species Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.