Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Myb, p Thr486 Antibody, Novus Biologicals™
SDP

Catalog No. NBP221659 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP221659 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP221659 Supplier Novus Biologicals Supplier No. NBP221659
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

c-Myb Polyclonal specifically detects c-Myb in Mouse samples. It is validated for Western Blot, ELISA.

Specifications

Antigen c-Myb (p Thr486)
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.91 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:2500, ELISA 1:5000-1:16000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. P06876
Gene Alias c-myb, Cmyb, c-myb protein (140 AA), c-myb_CDS, c-myb10A_CDS, c-myb13A_CDS, c-myb14A_CDS, c-myb8B_CDS, efg, Proto-oncogene c-Myb, transcriptional activator Myb, v-myb avian myeloblastosis viral oncogene homolog, v-myb myeloblastosis viral oncogene homolog (avian)
Gene Symbols MYB
Host Species Rabbit
Immunogen This affinity purified c-Myb [p Thr486] Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region of mouse c-Myb protein surrounding amino acid residue 486. (Uniprot: P06876)
Molecular Weight of Antigen 71.42 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 4602
Test Specificity This affinity-purified antibody is directed against the phosphorylated form of mouse c-Myb protein at the pT486 residue. The resultant affinity purified antibody was then cross adsorbed against the non-phosphorylated form of the immunizing peptide. Reactivity occurs against mouse c-Myb pT486 protein and the antibody is specific for the phosphorylated form of the protein. This antibody reacts with endogenous c-Myb protein and also SUMOylated overexpressed c-Myb protein in transfected Cos7 cells. Reactivity with non-phosphorylated mouse c-Myb is minimal by ELISA and western blot. A BLAST analysis was used to suggest reactivity with c-Myb from mouse based on a 100% homology with the immunizing sequence. Expect reactivity with c-Myb from rat, pig, human, horse, platypus, opossum, orangutan, spider monkey, bonobo, gorilla, and chimpanzee; with a v-myb myeloblastosis viral oncogene homolog, all isoforms (also avian version) from human; and with a protooncogene c-myb, and a v-myb myeloblastosis viral oncogene homolog (avian), both from cattle, all based on a 93% homology with the immunizing sequence. Cross-reactivity with c-Myb from other sources has not been determined.
Target Species Mouse
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.