Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Myc Antibody (2270A), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB371652 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB371652 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB371652 Supplier Novus Biologicals Supplier No. FAB36961I
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

c-Myc Monoclonal specifically detects c-Myc in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining), Gene Knock-Out, Immunocytochemistry, Western Blot.

Specifications

Antigen c-Myc
Applications CyTOF, Flow Cytometry (Intracellular Staining), Gene Knock-Out, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 2270A
Conjugate Janelia Fluor 549
Dilution Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready, Knockout Validated
Formulation 50mM Sodium Borate
Gene Alias avian myelocytomatosis viral oncogene homolog, BHLHE39, bHLHe39MRTL, Class E basic helix-loop-helix protein 39, c-Myc, MYC, myc proto-oncogene protein, MYCC, myc-related translation/localization regulatory factor, Proto-oncogene c-Myc, Transcription factor p64, v-myc avian myelocytomatosis viral oncogene homolog, v-myc myelocytomatosis viral oncogene homolog (avian)
Host Species Rabbit
Immunogen E.coli-derived recombinant human c-Myc, Arg66-Asp201, Accession # P01106
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cancer Stem Cells, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Epigenetics, Epitope Tags, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4609
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.