Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Myc Antibody (Myc.A7) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB397070 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397070 0.025 mg
NBP237822 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB397070 Supplier Novus Biologicals Supplier No. NBP2378220.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

c-Myc Monoclonal specifically detects c-Myc in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, CyTOF-ready.

Specifications

Antigen c-Myc
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, CyTOF
Classification Monoclonal
Clone Myc.A7
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000 - 1:3000, Flow Cytometry, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:500 - 1:2000, Immunoprecipitation 1:10 -1:500, CyTOF-ready
Formulation PBS (pH 7.2) with No Preservative
Gene Alias avian myelocytomatosis viral oncogene homolog, BHLHE39, bHLHe39MRTL, Class E basic helix-loop-helix protein 39, c-Myc, MYC, myc proto-oncogene protein, MYCC, myc-related translation/localization regulatory factor, Proto-oncogene c-Myc, Transcription factor p64, v-myc avian myelocytomatosis viral oncogene homolog, v-myc myelocytomatosis viral oncogene homolog (avian)
Gene Symbols MYC
Host Species Mouse
Immunogen Synthetic peptide: EQKLISEEDL c-Myc Antibody (Myc.A7).
Molecular Weight of Antigen 48.8 kDa
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cancer Stem Cells, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Epigenetics, Epitope Tags, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4609
Test Specificity Detects N-terminal or C-terminal Myc tagged fusion proteins. It has been successfully used in ELISA, immunofluorescence, immunoprecipitation and Western Blot procedures. immunogen is a synthetic peptide EQKLISEEDL (Myc).
Target Species Non-species specific
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.