Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
c-Myc Antibody (Myc.A7) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10 - $501.50
Specifications
Antigen | c-Myc |
---|---|
Clone | Myc.A7 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000 - 1:3000, Flow Cytometry, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:500 - 1:2000, Immunoprecipitation 1:10 -1:500, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, CyTOF |
Description
c-Myc Monoclonal specifically detects c-Myc in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, CyTOF-ready.Specifications
c-Myc | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, CyTOF | |
Unconjugated | |
Mouse | |
Autophagy, Cancer, Cancer Stem Cells, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Epigenetics, Epitope Tags, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors | |
PBS (pH 7.2) with No Preservative | |
4609 | |
Synthetic peptide: EQKLISEEDL c-Myc Antibody (Myc.A7). | |
Primary | |
Detects N-terminal or C-terminal Myc tagged fusion proteins. It has been successfully used in ELISA, immunofluorescence, immunoprecipitation and Western Blot procedures. immunogen is a synthetic peptide EQKLISEEDL (Myc). |
Myc.A7 | |
Western Blot 1:1000 - 1:3000, Flow Cytometry, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:500 - 1:2000, Immunoprecipitation 1:10 -1:500, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Non-species specific | |
avian myelocytomatosis viral oncogene homolog, BHLHE39, bHLHe39MRTL, Class E basic helix-loop-helix protein 39, c-Myc, MYC, myc proto-oncogene protein, MYCC, myc-related translation/localization regulatory factor, Proto-oncogene c-Myc, Transcription factor p64, v-myc avian myelocytomatosis viral oncogene homolog, v-myc myelocytomatosis viral oncogene homolog (avian) | |
MYC | |
IgG1 | |
Protein A purified | |
48.8 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title