Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calreticulin Antibody, Novus Biologicals™
SDP

Catalog No. NB600103 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600103 0.1 mL
NB600103SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB600103 Supplier Novus Biologicals Supplier No. NB600103
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 5 publications

Calreticulin Polyclonal specifically detects Calreticulin in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated, Knockdown Validated.

Specifications

Antigen Calreticulin
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay, KnockDown
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000, Simple Western 1:500, Immunohistochemistry 1:20, Immunocytochemistry/Immunofluorescence 1:20-1:100. Use reported by customer review, Immunohistochemistry-Paraffin 1:20, Immunohistochemistry-Frozen reported in scientific literature (PMID 26643212), Knockout Validated, KnockDown Validated 1:2500
Formulation Whole antisera with 0.1% Sodium Azide
Gene Accession No. P14211
Gene Alias autoantigen Ro, calregulin, calreticulin, cC1qR, CRP55, CRT, CRTC, Endoplasmic reticulum resident protein 60, ERp60, FLJ26680, grp60, RO, Sicca syndrome antigen A (autoantigen Ro; calreticulin), SSAHACBP
Gene Symbols CALR
Host Species Rabbit
Immunogen Calreticulin Antibody was developed against a full-length protein to mouse Calreticulin [UniProt# P14211]
Molecular Weight of Antigen 48 kDa
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cellular Markers, ER Markers, Immunology, Membrane Trafficking and Chaperones, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 811
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.