Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Myc Antibody (SPM237), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB120961 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120961 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB120961 Supplier Novus Biologicals Supplier No. NBP286682JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

c-Myc Monoclonal antibody specifically detects c-Myc in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence

Specifications

Antigen c-Myc
Applications Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone SPM237
Conjugate Janelia Fluor 549
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias avian myelocytomatosis viral oncogene homolog, BHLHE39, bHLHe39MRTL, Class E basic helix-loop-helix protein 39, c-Myc, MYC, myc proto-oncogene protein, MYCC, myc-related translation/localization regulatory factor, Proto-oncogene c-Myc, Transcription factor p64, v-myc avian myelocytomatosis viral oncogene homolog, v-myc myelocytomatosis viral oncogene homolog (avian)
Host Species Mouse
Immunogen A synthetic peptide, corresponding to aa 408-439 (AEEQKLISEEDLLRKRREQLKHKLEQLRNSCA) from C-terminus of human c-Myc, coupled to KLH.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cancer Stem Cells, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Epigenetics, Epitope Tags, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4609
Target Species Human, Chicken (Negative), Mouse (Negative), Rat (Negative)
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.