Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Myc, p Ser62 Antibody (2A10), Novus Biologicals™
SDP

Catalog No. NB298169 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298169 100 μL
NB298168 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298169 Supplier Novus Biologicals Supplier No. NBP326404100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

c-Myc Monoclonal antibody specifically detects c-Myc in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry

Specifications

Antigen c-Myc
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 2A10
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias avian myelocytomatosis viral oncogene homolog, BHLHE39, bHLHe39MRTL, Class E basic helix-loop-helix protein 39, c-Myc, MYC, myc proto-oncogene protein, MYCC, myc-related translation/localization regulatory factor, Proto-oncogene c-Myc, Transcription factor p64, v-myc avian myelocytomatosis viral oncogene homolog, v-myc myelocytomatosis viral oncogene homolog (avian)
Host Species Rabbit
Immunogen A synthesized peptide derived from Human c-Myc [p Ser62] [UniProt P01106]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cancer Stem Cells, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Epigenetics, Epitope Tags, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4609
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.