Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Myc-responsive protein Rcl Rabbit anti-Human, Mouse, Clone: 7C1Z8, Novus Biologicals™
SDP

Catalog No. NB167321 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167321 100 μg
NB167322 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB167321 Supplier Novus Biologicals Supplier No. NBP316115100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

c-Myc-responsive protein Rcl Monoclonal antibody specifically detects c-Myc-responsive protein Rcl in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen c-Myc-responsive protein Rcl
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 7C1Z8
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias chromosome 6 open reading frame 108, c-Myc-responsive protein Rcl, deoxyribonucleoside 5'-monophosphate N-glycosidase, dJ330M21.3, EC 3.2.2, EC 3.2.2.-, putative c-Myc-responsive, rcl, RP3-330M21.3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 75-174 of human c-Myc-responsive protein Rcl (O43598).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 10591
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.