Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C-Peptide Antibody (2B7), Novus Biologicals™
SDP

Catalog No. NB1102555 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1102555 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1102555 Supplier Novus Biologicals Supplier No. NB1102555

Mouse Monoclonal Antibody

C-Peptide Monoclonal specifically detects C-Peptide in Human, Bovine (Negative), Mouse (Negative), Porcine (Negative), Rat (Negative) samples. It is validated for ELISA, Radioimmunodiffusion, Radioimmunoassay, Competitive ELISA.

Specifications

Antigen C-Peptide
Applications ELISA, Radioimmune Assays (RIA)
Classification Monoclonal
Clone 2B7
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Radioimmunodiffusion, Radioimmunoassay 1:100-1:2000, Competitive ELISA
Gene Alias IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
Gene Symbols INS
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of X63-Ag8-653 myeloma cells with spleen cells of Balb/c mice immunized with recombinant C-peptide construction.
Purification Method Protein A purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Diabetes Research
Primary or Secondary Primary
Gene ID (Entrez) 3630
Test Specificity Affinity (Kd) > 1 x 10^8. Binds to the human free C-peptide and C-peptide region in proinsulin molecules. Antigenic determinants identified by MAbs are in the three-dimensional structure generated by N- and C- terminal regions of C-peptide separated by beta-turn at position 47-50 of proinsulin.. No cross-reactivity with human, bovine, porcine and mouse/rat insulin.
Target Species Human, Bovine (Negative), Mouse (Negative), Porcine (Negative), Rat (Negative)
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.