Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C-Peptide Antibody (CC34), Novus Biologicals™
SDP

Catalog No. NBP105433 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP105433 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP105433 Supplier Novus Biologicals Supplier No. NBP105433
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C-Peptide Monoclonal specifically detects C-Peptide in Mouse, Rat samples. It is validated for ELISA, Immunoassay, Immunohistochemistry, Immunohistochemistry (Frozen), Sandwich ELISA.

Specifications

Antigen C-Peptide
Applications ELISA, Immunoassay, Immunohistochemistry, Immunohistochemistry (Frozen), Sandwich ELISA
Classification Monoclonal
Clone CC34
Conjugate Unconjugated
Dilution ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Immunoassay, Sandwich ELISA 1:100 - 1:2000
Gene Alias IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
Gene Symbols INS
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with fragments of rat C-peptides I and II conjugated with the carrier protein.
Purification Method Protein A purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Diabetes Research
Primary or Secondary Primary
Gene ID (Entrez) 3630
Test Specificity Reacts with rat c-peptide I, rat c-peptide II, mouse c-peptide II and mouse c-peptide I. Does not react with rat proinsulin.
Target Species Mouse, Rat
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.