Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C-Peptide Rabbit anti-Human, Clone: SA0410, Novus Biologicals™
SDP

Catalog No. NBP267288 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267288 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267288 Supplier Novus Biologicals Supplier No. NBP267288

Rabbit Monoclonal Antibody

C-Peptide Monoclonal specifically detects C-Peptide in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, ICC/IF (Negative).

Specifications

Antigen C-Peptide
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SA0410
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:200-1:500, Immunohistochemistry-Paraffin 1:20000
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
Gene Symbols INS
Host Species Rabbit
Immunogen Recombinant protein within human C-Peptide aa 15-110. (SwissProt: P01308 Human; SwissProt: P01325 Mouse; SwissProt: P01322 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Diabetes Research, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 3630
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.