Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C-Reactive Protein/CRP Antibody (5A9), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP104282A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP104282A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP104282A94 Supplier Novus Biologicals Supplier No. NBP104282AF594

Mouse Monoclonal Antibody

C-Reactive Protein/CRP Monoclonal specifically detects C-Reactive Protein/CRP in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen C-Reactive Protein/CRP
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 5A9
Conjugate Alexa Fluor 594
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias C-reactive protein, C-reactive protein, pentraxin-related, MGC88244, pentraxin 1, PTX1MGC149895
Gene Symbols CRP
Host Species Mouse
Immunogen Recombinant human C-Reactive Protein (19-224aa) purified from E. coli
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 1401
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.