Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C-Reactive Protein/CRP Antibody (C1), HRP
SDP

Catalog No. NB12010026H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB12010026H 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB12010026H Supplier Novus Biologicals Supplier No. NB12010026H

Mouse Monoclonal Antibody

C-Reactive Protein/CRP Monoclonal specifically detects C-Reactive Protein/CRP in Human samples. It is validated for Western Blot.

Specifications

Antigen C-Reactive Protein/CRP
Applications Western Blot
Classification Monoclonal
Clone C1
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Alias C-reactive protein, C-reactive protein, pentraxin-related, MGC88244, pentraxin 1, PTX1MGC149895
Gene Symbols CRP
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Human CRP.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Cancer, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 1401
Test Specificity does not react with Ca2+ depleted protein
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.