Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C-Reactive Protein/CRP Antibody (C7), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB359097 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359097 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359097 Supplier Novus Biologicals Supplier No. NB6001333AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C-Reactive Protein/CRP Monoclonal antibody specifically detects C-Reactive Protein/CRP in Human samples. It is validated for ELISA, Immunohistochemistry, Immunoassay, ELISA, Affinity Purification

Specifications

Antigen C-Reactive Protein/CRP
Applications ELISA, Immunohistochemistry, Immunoassay, ELISA, Affinity Purification
Classification Monoclonal
Clone C7
Conjugate Alexa Fluor 532
Dilution ELISA, Immunohistochemistry, Immunoassay, Sandwich ELISA, Immunoaffinity Purification
Formulation 50mM Sodium Borate
Gene Alias C-reactive protein, C-reactive protein, pentraxin-related, MGC88244, pentraxin 1, PTX1MGC149895
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Human CRP.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Cancer, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 1401
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.