Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C12orf49 Antibody, Novus Biologicals™
SDP

Catalog No. NBP16250820 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP16250820 20 μL
NBP162508 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP16250820 Supplier Novus Biologicals Supplier No. NBP16250820UL

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

C12orf49 Polyclonal specifically detects C12orf49 in Human samples. It is validated for Western Blot.

Specifications

Antigen C12orf49
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9H741
Gene Alias chromosome 12 open reading frame 49, FLJ21415, hypothetical protein LOC79794
Gene Symbols C12ORF49
Host Species Rabbit
Immunogen Synthetic peptides corresponding to C12ORF49 The peptide sequence was selected from the C terminal of C12ORF49. Peptide sequence LERFLNRAAVAFQNLFMAVEDHFELCLAKCRTSSQSVQHENTYRDPIAKY.
Molecular Weight of Antigen 23 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79794
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.