Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C14orf180 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | C14orf180 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15953320
![]() |
Novus Biologicals
NBP15953320UL |
20 μL |
Each for $206.00
|
|
|||||
NBP159533
![]() |
Novus Biologicals
NBP159533 |
100 μL |
Each for $487.50
|
|
|||||
Description
C14orf180 Polyclonal specifically detects C14orf180 in Human samples. It is validated for Western Blot.Specifications
C14orf180 | |
Polyclonal | |
Rabbit | |
Human | |
C14orf77, chromosome 14 open reading frame 180, chromosome 14 open reading frame 77, FLJ38594, NRAC, transmembrane protein C14orf180 | |
C14ORF180 | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
Q8N912 | |
400258 | |
Synthetic peptides corresponding to C14ORF180 The peptide sequence was selected from the N terminal of C14ORF180. Peptide sequence EDNRKCPPSILKRSRPEHHRPEAKPQRTSRRVWFREPPAVTVHYIADKNA. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title