Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C15orf40 Antibody (OTI2B7), PE/Cy7, Novus Biologicals™
SDP

Catalog No. N272375PEC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272375PEC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272375PEC7 Supplier Novus Biologicals Supplier No. NBP272375PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C15orf40 Monoclonal specifically detects C15orf40 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen C15orf40
Applications Flow Cytometry
Classification Monoclonal
Clone OTI2B7
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias chromosome 15 open reading frame 40, FLJ33606, hypothetical protein LOC123207, MGC29937
Gene Symbols C15orf40
Host Species Mouse
Immunogen Full length recombinant protein of human C15orf40 (NP_653198) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 123207.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.