Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C16orf72 Antibody (OTI2D1), Biotin, Novus Biologicals™
SDP

Catalog No. NBP271842B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271842B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271842B Supplier Novus Biologicals Supplier No. NBP271842B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C16orf72 Monoclonal specifically detects C16orf72 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen C16orf72
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI2D1
Conjugate Biotin
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias chromosome 16 open reading frame 72, FLJ41272, hypothetical protein LOC29035, PRO0149
Gene Symbols C16orf72
Host Species Mouse
Immunogen Full length recombinant protein of human C16orf72 (NP_054836) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 29035.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.