Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C17orf28 Monoclonal Antibody (OTI3B5), TrueMAB™, OriGene
SDP

Catalog No. 502542352
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2352 30 μL
50-167-2502 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2352 Supplier OriGene Supplier No. TA501359S
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C17orf28 Monoclonal antibody specifically detects C17orf28 in Human, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen C17orf28
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI3B5
Concentration 0.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene TBXAS1
Gene Accession No. P24557, P49430
Gene Alias C17orf28; C630004H02Rik; C9H17orf28; DMC1; downregulated in multiple cancer 1; Down-regulated in multiple cancers 1; Hid1; HID-1; HID1 domain containing; HID1 domain-containing protein; protein HID1; Protein hid-1 homolog; RGD1311422; UPF0663 transmembrane protein C17orf28; UPF0663 transmembrane protein C17orf28 homolog
Host Species Mouse
Immunogen Full length human recombinant protein of human TBXAS1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24886, 6916
Target Species Human, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.