Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C17orf58 Antibody (OTI6E9), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP272489A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP272489A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP272489A32 Supplier Novus Biologicals Supplier No. NBP272489AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C17orf58 Monoclonal specifically detects C17orf58 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen C17orf58
Applications Western Blot
Classification Monoclonal
Clone OTI6E9
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias chromosome 17 open reading frame 58, hypothetical protein LOC284018, MGC138278
Gene Symbols C17orf58
Host Species Mouse
Immunogen Full length recombinant protein of human C17orf58 (NP_858041) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 284018.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.