Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C17orf58 Antibody (OTI6E9), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB288112 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288112 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288112 Supplier Novus Biologicals Supplier No. NBP272489APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C17orf58 Monoclonal antibody specifically detects C17orf58 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen C17orf58
Applications Western Blot
Classification Monoclonal
Clone OTI6E9
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias chromosome 17 open reading frame 58, hypothetical protein LOC284018, MGC138278
Host Species Mouse
Immunogen Full length human recombinant protein of human C17orf58(NP_858041) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284018
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.