Learn More
Description
Specifications
Specifications
| Antigen | C19orf47 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000 |
| Formulation | PBS and 2% Sucrose with 0.09% Sodium Azide |
| Gene Alias | chromosome 19 open reading frame 47, DKFZp686P05129, FLJ36888, hypothetical protein LOC126526 |
| Gene Symbols | C19ORF47 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptides corresponding to C19ORF47 The peptide sequence was selected from the middle region of C19ORF47. Peptide sequence YVINMPKGTTPRTRKILEQQQAAKGLHRTSVFDRLGAETKADTTTGSKPT. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
