Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1D Antibody (004), FITC, Novus Biologicals™
SDP

Catalog No. NB334627 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB334627 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB334627 Supplier Novus Biologicals Supplier No. NBP290103F
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

C1D Monoclonal antibody specifically detects C1D in Human samples. It is validated for Western Blot

Specifications

Antigen C1D
Applications Western Blot
Classification Monoclonal
Clone 004
Conjugate FITC
Dilution Western Blot
Formulation PBS
Gene Alias C1D DNA-binding protein, C1D nuclear receptor corepressor, C1D nuclear receptor co-repressor, hC1D, MGC12261, MGC14659, nuclear DNA-binding protein, nuclear nucleic acid-binding protein C1D, small unique nuclear receptor corepressor, small unique nuclear receptor co-repressor, SUN-CoR, SUNCORLRP1
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Human C1D (Uniprot#: Q13901, Met1-Ser141)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 10438
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.