Learn More
Description
Specifications
Specifications
| Antigen | C1D |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 42 |
| Conjugate | Alexa Fluor 750 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | C1D DNA-binding protein, C1D nuclear receptor corepressor, C1D nuclear receptor co-repressor, hC1D, MGC12261, MGC14659, nuclear DNA-binding protein, nuclear nucleic acid-binding protein C1D, small unique nuclear receptor corepressor, small unique nuclear receptor co-repressor, SUN-CoR, SUNCORLRP1 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human C1D (Q13901; Met1-Ser141). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
