Learn More
Description
Specifications
Specifications
| Antigen | C1orf105 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | chromosome 1 open reading frame 105, hypothetical protein LOC92346, RP1-106H8.1 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human C1orf105 (NP_640333). Peptide sequence PEPFHDDIPTESIHYRLPILGPRTAVFHGLLTEAYKTLKERQRSSLPRKE |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
