Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1orf50 Antibody (OTI1C6), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N27218APCY7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N27218APCY7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N27218APCY7 Supplier Novus Biologicals Supplier No. NBP272188APCCY7

Mouse Monoclonal Antibody

C1orf50 Monoclonal specifically detects C1orf50 in Human, Mouse, Canine, Primate, Monkey samples. It is validated for Flow Cytometry.

Specifications

Antigen C1orf50
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1C6
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias chromosome 1 open reading frame 50, hypothetical protein LOC79078, MGC955
Gene Symbols C1orf50
Host Species Mouse
Immunogen Full length recombinant protein of human C1orf50 (NP_077002) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 79078.0
Target Species Human, Mouse, Canine, Primate, Monkey
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.