Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1QB Antibody (C1QB/2961), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB012600 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB012600 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB012600 Supplier Novus Biologicals Supplier No. NBP279920V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C1QB Monoclonal specifically detects C1QB in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen C1QB
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone C1QB/2961
Conjugate DyLight 405
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias B chain, complement C1q subcomponent subunit B, complement component 1, q subcomponent, B chain, complement subcomponent C1q chain B, q subcomponent, beta polypeptide
Gene Symbols C1QB
Host Species Mouse
Immunogen Recombinant fragment (around aa 41-188) of human C1QB protein (exact sequence is proprietary) (Uniprot: P02746)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.