Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1QB Antibody (C1QB/2962), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB156417 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB156417 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB156417 Supplier Novus Biologicals Supplier No. NBP314195FR
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

C1QB Monoclonal antibody specifically detects C1QB in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen C1QB
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone C1QB/2962
Conjugate DyLight 680
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias B chain, complement C1q subcomponent subunit B, complement component 1, q subcomponent, B chain, complement subcomponent C1q chain B, q subcomponent, beta polypeptide
Host Species Mouse
Immunogen Recombinant fragment (around aa 41-188) of human C1QB protein (exact sequence is proprietary) (Uniprot: P02746 )
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Immunology, Innate Immunity, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 713
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.