Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1QL4 Rabbit anti-Human, Polyclonal, Invitrogen™

Catalog No. PA548224
Change view
Click to view available options
Quantity:
400 μL
Catalog No. Quantity
PA548224 400 μL
1 options

Catalog No. PA548224

Supplier: Thermo Scientific PA548224

Rabbit Polyclonal Antibody

May regulate the number of excitatory synapses that are formed on hippocampus neurons. Has no effect on inhibitory synapses. May inhibit adipocyte differentiation at an early stage of the process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1QL4
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene C1QL4
Gene Accession No. Q86Z23
Gene Alias Complement C1q-like protein 4, C1q and tumor necrosis factor-related protein 11, C1q/TNF-related protein 11, C1QL4, CTRP11
Gene Symbols C1QL4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human C1QL4.
Purification Method Antigen affinity chromatography
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338761
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.