Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1qR1/CD93 Antibody (004), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB070182 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB070182 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB070182 Supplier Novus Biologicals Supplier No. NBP290128UV
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

C1qR1/CD93 Monoclonal specifically detects C1qR1/CD93 in Human samples. It is validated for ELISA.

Specifications

Antigen C1qR1/CD93
Applications ELISA
Classification Monoclonal
Clone 4
Conjugate DyLight 350
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias C1q receptor 1, C1q/MBL/SPA receptor, C1qR(P), C1qRp, CD93 antigenC1qR, CD93 molecule, CDw93C1QR1, Complement component 1 q subcomponent receptor 1, complement component 1, q subcomponent, receptor 1, complement component C1q receptor, dJ737E23.1, ECSM3, Matrix-remodeling-associated protein 4, matrix-remodelling associated 4, MXRA4
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human C1qR1/CD93 (Q9NPY3; Met1-Lys580).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers
Primary or Secondary Primary
Gene ID (Entrez) 22918
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.