Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1qR1/CD93 Antibody (164), PerCP, Novus Biologicals™
SDP

Catalog No. NB315882 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB315882 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB315882 Supplier Novus Biologicals Supplier No. NBP290894PCP
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

C1qR1/CD93 Monoclonal antibody specifically detects C1qR1/CD93 in Rat samples. It is validated for ELISA

Specifications

Antigen C1qR1/CD93
Applications ELISA
Classification Monoclonal
Clone 164
Conjugate PerCP
Dilution ELISA
Formulation PBS
Gene Alias C1q receptor 1, C1q/MBL/SPA receptor, C1qR(P), C1qRp, CD93 antigenC1qR, CD93 molecule, CDw93C1QR1, Complement component 1 q subcomponent receptor 1, complement component 1, q subcomponent, receptor 1, complement component C1q receptor, dJ737E23.1, ECSM3, Matrix-remodeling-associated protein 4, matrix-remodelling associated 4, MXRA4
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Rat C1qR1/CD93 (Uniprot#: Q9ET61; Met1-Lys571).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers
Primary or Secondary Primary
Gene ID (Entrez) 22918
Target Species Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.