Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1qTNF5/CTRP5 Antibody (332923), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB348494 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB348494 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB348494 Supplier Novus Biologicals Supplier No. FAB3167MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C1qTNF5/CTRP5 Monoclonal antibody specifically detects C1qTNF5/CTRP5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry, CyTOF

Specifications

Antigen C1qTNF5/CTRP5
Applications Western Blot, Immunohistochemistry, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 332923
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias C1q and tumor necrosis factor related protein 5, Complement C1q Tumor Necrosis Factor-Related Protein 5 Precursor Variant 3, CTRP5LORDDKFZp586B0621, LORD 6, Myonectin 3
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CTRP5/C1qTNF5, Ser16-Ala243 (Gln44Arg), Accession # Q9BXJ0
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 114902
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.