Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C21orf59 Antibody (OTI2H5), PE/Cy7, Novus Biologicals™
SDP

Catalog No. N272060PEC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272060PEC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272060PEC7 Supplier Novus Biologicals Supplier No. NBP272060PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C21orf59 Monoclonal specifically detects C21orf59 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen C21orf59
Applications Flow Cytometry
Classification Monoclonal
Clone OTI2H5
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias C21orf48, chromosome 21 open reading frame 48, chromosome 21 open reading frame 59, FLJ20467, FLJ37137, FLJ40247, hypothetical protein LOC56683, kur, kurly
Gene Symbols CFAP298
Host Species Mouse
Immunogen Full length recombinant protein of human C21orf59 (NP_067077) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 56683.0
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.