Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ C28

Click to view available options
Quantity:
10 mg
50 mg
Description
C28 is a potent lemur tyrosine kinase 3 (LMTK3) inhibitor (IC50 = 67 nM). Disrupts interaction of LMTK3 and HSP90 leading to degradation of LMTK3. Also inhibits CLK, DYRK1α, IRAK4 and HIPK2 (IC50 values are 5 nM, 6 nM, 41 nM and 48 nM respectively). C28 inhibits growth and increases apoptosis in breast cancer cell lines (IC50 = 6.5-14.77 μM) and reduces growth of tumors in xenograft and transgenic mouse models of breast cancer. Also reduces levels of ERα.
Specifications
Specifications
Chemical Name or Material | 6-(Pyridin-4-yl)-3-(3-(trifluoromethoxy)phenyl)imidazo[1,2-b]pyridazine |
Quantity | 10 mg |
Target | LMTK3 |
Molecular Formula | C18H11F3N4O |
Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction