Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ C2C12 Whole Cell Lysate
SDP

Catalog No. NBP21026801 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP21026801 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP21026801 Supplier Novus Biologicals™ Supplier No. NBP2102680.1MG

C2C12 (muscle, myoblast) whole stem cell lysate 2mg/mL supplied in SDS sample buffer containing 5% Beta-mercaptoethanol. Growth media: DMEM and 10% FBS (fetal bovine serum). Lysates are guaranteed for 6 months from date of receipt.

The C2C12 Whole Cell Lysate is derived from mouse. It has been validated for the following applications: Western Blot.

Specifications

Tissue C2C12
For Use With (Application) Western Blot
Quantity 0.1 mg
Storage Buffer Supplied in SDS sample buffer containing 5% Beta-mercaptoethanol.
Test Specificity C2C12 (muscle; myoblast)
Host Species Mouse
Concentration 2 mg/mL
Content And Storage Store at −80°C. Avoid freeze-thaw cycles.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.