Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C2CD3 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017274 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017274 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB017274 Supplier Novus Biologicals Supplier No. NBP281963
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C2CD3 Polyclonal specifically detects C2CD3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.

Specifications

Antigen C2CD3
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA
Formulation PBS
Gene Alias C2 calcium-dependent domain containing 3
Gene Symbols C2CD3
Host Species Rabbit
Immunogen C2CD3 antibody was raised against an 18 amino acid peptide near the center of human C2CD3. The immunogen is located within amino acids 1490 - 1540 of C2CD3. Amino Acid Squence: YGNDSVERPHQTDSWIGS
Molecular Weight of Antigen 255 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26005
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.