Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C2CD5 Antibody, Novus Biologicals™
SDP

Catalog No. NB10074595 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10074595 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10074595 Supplier Novus Biologicals Supplier No. NB10074595
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KIAA0528 Polyclonal specifically detects KIAA0528 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).

Specifications

Antigen KIAA0528
Applications Immunoprecipitation, Western Blot (Negative)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 2-5 ug/mg lysate, Western Blot (Negative)
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_055617.1
Gene Alias DKFZp779N2044, hypothetical protein LOC9847, KIAA0528
Gene Symbols C2CD5
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 250 and 300 of human KIAA0528 using the numbering given in entry NP_055617.1 (GeneID 9847).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9847
Test Specificity Human. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Chicken, Turkey, Bovine, Dog, Horse, Rabbit, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, European domestic ferret, African elephant, Naked mole rat, Gray short-tailed opossum and Northern white-cheeked gibbon.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.