Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C2orf88 Antibody, Novus Biologicals™
SDP

Catalog No. NB186603 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB186603 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB186603 Supplier Novus Biologicals Supplier No. NBP312454
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

C2orf88 Polyclonal antibody specifically detects C2orf88 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen C2orf88
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Formulation Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide
Gene Alias chromosome 2 open reading frame 88, hypothetical protein LOC84281, MGC13057
Host Species Rabbit
Immunogen N-epitope synthetic peptide taken within amino acid region 1-50 on human C2orf88 protein.
Purification Method Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84281
Target Species Human
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.