Learn More
Tocris Bioscience™ C34
GREENER_CHOICE
Catalog No. p-5546142
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
Catalog No. 53-731-0
Supplier: Tocris Bioscience™ 5373/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
TLR4 inhibitor
C34 is a TLR4 inhibitor; attenuates TLR4 signaling in enterocytes and macrophages in vitro, and reduces systemic inflammation in mouse models of endotoxemia and necrotizing enterocolitis (NEC). Also suppresses LPS signaling in human ileum displaying NEC ex-vivo. Displays no significant effects on signaling via TLR2 or TLR9.
Specifications
Quantity 10 mg
Product Type C34
Molecular Weight (g/mol) 389.4
Inhibitors C34
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.