Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C3orf36 Antibody (OTI8G4), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. N272218A405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272218A405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272218A405 Supplier Novus Biologicals Supplier No. NBP272218AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C3orf36 Monoclonal specifically detects C3orf36 in Human samples. It is validated for Western Blot.

Specifications

Antigen C3orf36
Applications Western Blot
Classification Monoclonal
Clone OTI8G4
Conjugate Alexa Fluor 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C3orf36 chromosome 3 open reading frame 36
Gene Symbols C3orf36
Host Species Mouse
Immunogen Full length recombinant protein of human C3orf36 (NP_079317) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 80111.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.