Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C4 binding protein B Antibody (SR1762), Novus Biologicals™
SDP

Catalog No. NB259222 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB259222 100 μg
NB260090 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB259222 Supplier Novus Biologicals Supplier No. NBP321995100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

C4 binding protein B Monoclonal antibody specifically detects C4 binding protein B in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation

Specifications

Antigen C4 binding protein B
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone SR1762
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:60
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias C4b binding protein, beta chain, C4b-binding protein beta chain, C4BP, complement component 4 binding protein, beta, complement component 4 binding protein, beta chain, complement component 4-binding protein, beta
Host Species Rabbit
Immunogen A synthesized peptide derived from human C4 binding protein B (Uniprot #: P20851)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 725
Target Species Human, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.