Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C4orf14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598671
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598671 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598671 Supplier Invitrogen™ Supplier No. PA598671
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C4orf14 Polyclonal Antibody for Western Blot, IHC (P), ELISA

The protein encoded by this gene is a nuclear-encoded GTPase that functions in the mitochondrion. Upon translation, this protein is imported into the nucleus and then into the nucleolus before being exported to the mitochondrion. The encoded protein is required for oxygen-dependent regulation of mitochondrial respiratory complexes and for mitochondrial protein synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C4orf14
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene NOA1
Gene Accession No. Q8NC60, Q9JJG9
Gene Alias 2610024G14Rik; AtNOS1; AU022345; C4orf14; hAtNOS1; hNOA1; mAtNOS1; mitochondrial GTPase 3 homolog; MMR_HSR1 domain containing protein RGD1359460; MNCb-4931; MTG3; nitric oxide associated 1; nitric oxide associated-1; nitric oxide synthase, mitochondrial (putative); nitric oxide-associated protein 1; Noa1; putative ortholog of Arabidopsis mitochondrial nitric oxide synthase; RGD1359460
Gene Symbols NOA1
Host Species Rabbit
Immunogen Recombinant Human Nitric oxide-associated protein 1 protein (42-174AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56412, 84273
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.