Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C5 Mouse anti-Human, Clone: 2G8C9, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | C5 |
---|---|
Clone | 2G8C9 |
Concentration | 0.93 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7052
![]() ![]() |
Proteintech Group Inc
666341IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
C5 is part of the membrane attack complex (MAC) of the complement system. C5 is split into C5a og C5b by the C5 convertases from the classical and the alternative complement pathway and the C5b fragment initiates the assembly of the terminal MAC (C5b-C9). The molecular mass of C5 is 190 kDa.Specifications
C5 | |
0.93 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C5, Complement C5, complement component 5, CPAMD4 | |
C5 | |
IgG2a | |
Protein A | |
Antibody |
2G8C9 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P01031 | |
727 | |
C5 Fusion Protein Ag17882 | |
Primary | |
-20°C | |
C5 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title